Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sec5
Entrez Gene ID
33563
Gene Name
Secretory 5
Gene Aliases
CG8843, Dmel\CG8843, Dmel_CG8843, SEC5, Sec-5, Sec5p, dsec5, sec5
Chromosome Location
Chr.2L: 3457302 - 3460367 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266670
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134936.3 NP_608780.1 1 - 2 220 81
GenBank mRNA AY069793.1 - 1 - 2 180 81

View Full Product Details