Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGRP-LC
Entrez Gene ID
39063
Gene Name
Peptidoglycan recognition protein LC
Gene Aliases
(PGRP)-LC, CG4432, Dm PGRP-LC, Dmel\CG4432, Dmel_CG4432, PGRP-LCa, PGRP-LCd, PGRP-LCx, PGRPLC, Pgrp-LC, ird7, pgrp lc, pgrp-lc, tot
Chromosome Location
Chr.3L: 9338810 - 9348336 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035976
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259764.1 NP_001246693.1 3 - 4 1241 96
NM_140041.4 NP_648298.1 3 - 4 1345 96
GenBank mRNA AF207539.1 - 3 - 4 1325 96
AY119048.1 - 3 - 4 1325 96

View Full Product Details