Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PGRP-LF
- Entrez Gene ID
-
39064
- Gene Name
- Peptidoglycan recognition protein LF
- Gene Aliases
-
CG4437, Dmel\CG4437, Dmel_CG4437, PGRP-like, PGRPLF, anon-WO0118547.581, pgrp-lf
- Chromosome Location
-
Chr.3L: 9349609 - 9351489 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035977
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140042.3 |
NP_648299.3 |
3 - 4 |
753 |
75 |
GenBank mRNA |
AY084190.1 |
- |
3 - 4 |
751 |
75 |
View Full Product Details