Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tna
Entrez Gene ID
39217
Gene Name
tonalli
Gene Aliases
0583/02, CG7958, Dmel\CG7958, Dmel_CG7958, anon-WO0118547.590, l(3)L6731, l(3)S058302, l(3)rI075
Chromosome Location
Chr.3L: 10835528 - 10876496 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026160
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169942.2 NP_001163413.1 3 - 4 1373 75
NM_001382194.1 NP_001368945.1 3 - 4 940 75
NM_140155.4 NP_648412.2 3 - 4 1520 75
NM_168422.3 NP_729629.2 2 - 3 935 75
GenBank mRNA BT003272.1 - 3 - 4 1373 75

View Full Product Details