Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sprn
Entrez Gene ID
39344
Gene Name
Spermitin
Gene Aliases
CG14128, Dmel\CG14128, Dmel_CG14128, sprn
Chromosome Location
Chr.3L: 11879115 - 11881182 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036218
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169953.1 NP_001163424.1 2 - 3 934 132
NM_001169954.2 NP_001163425.2 2 - 3 986 132
NM_001169955.1 NP_001163426.1 2 - 3 934 132
NM_140262.2 NP_648519.2 1 - 2 988 132
GenBank mRNA BT022853.1 - 2 - 3 927 132
BT022877.1 - 2 - 3 934 132

View Full Product Details