Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tral
Entrez Gene ID
39430
Gene Name
trailer hitch
Gene Aliases
BEST:GH08269, CG10686, DmLSm15, Dmel\CG10686, Dmel_CG10686, LSm15, Lsm15, Scd6p, TRAL, Tral, anon-EST:Posey139, anon-WO0172774.89, anon-WO02059370.24, p67, p70
Chromosome Location
Chr.3L: 12515059 - 12520954 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041775
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274840.1 NP_001261769.1 2 - 3 1009 87
NM_001274843.1 NP_001261772.1 2 - 3 1009 87
NM_001274844.1 NP_001261773.1 2 - 3 1009 87
NM_001274845.1 NP_001261774.1 2 - 3 1009 87
NM_140328.5 NP_648585.3 2 - 3 1009 87
GenBank mRNA AY069066.1 - 2 - 3 991 87

View Full Product Details