The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144473.3 | NP_001137945.1 | 6 - 7 | 1262 | 74 |
NM_001274856.2 | NP_001261785.1 | 4 - 5 | 802 | 74 |