Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
chic
Entrez Gene ID
33834
Gene Name
chickadee
Gene Aliases
CG9553, Chi, Chic, ChicE1, D88, Dmel\CG9553, Dmel_CG9553, GH4, chi, l(2)27/7, profilin, sand
Chromosome Location
Chr.2L: 5972115 - 5981018 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000308
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258976.2 NP_001245905.1 3 - 4 636 60
NM_001258977.2 NP_001245906.1 3 - 4 606 60
NM_057668.6 NP_477016.1 3 - 4 532 60
NM_134304.3 NP_599131.1 3 - 4 626 60
NM_164667.2 NP_723136.1 3 - 4 737 60
NM_205913.3 NP_995635.1 3 - 4 530 60
GenBank mRNA AY069444.1 - 2 - 3 582 60
AY128443.1 - 3 - 4 539 60
DQ062770.1 - 2 - 3 282 60
KX531778.1 - 2 - 3 282 60
M84528.1 - 3 - 4 623 60
M84529.1 - 3 - 4 496 60

View Full Product Details