Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nhe3
Entrez Gene ID
33939
Gene Name
Na[+]/H[+] hydrogen exchanger 3
Gene Aliases
CG11328, DmNHE3, Dmel\CG11328, Dmel_CG11328, NHE3, nhe3
Chromosome Location
Chr.2L: 6677835 - 6684364 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028703
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103635.3 NP_001097105.3 1 - 2 640 62
NM_001144329.2 NP_001137801.1 1 - 2 633 62
NM_001273218.1 NP_001260147.1 1 - 2 633 62
NM_135220.3 NP_609064.3 1 - 2 633 62
NM_164701.2 NP_723202.2 1 - 2 633 62

View Full Product Details