Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Piezo
Entrez Gene ID
34112
Gene Name
CG44122-PE
Gene Aliases
CG18103, CG31608, CG44122, CG8486, DmPIEZO, DmPiezo, Dmel\CG44122, Dmel_CG44122, Dmpiezo, OrfKD, anon-EST:Liang-1.32, clone 1.32, fos28F, fos500, piezo
Chromosome Location
Chr.2L: 8163073 - 8190137 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264953
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042881.5 NP_001036346.3 12 - 13 3458 68
NM_001169436.3 NP_001162907.1 11 - 12 3236 68
NM_001201789.2 NP_001188718.1 12 - 13 3293 68
NM_001201790.2 NP_001188719.1 11 - 12 3236 68
NM_001273297.1 NP_001260226.1 13 - 14 3515 68
NM_001273298.1 NP_001260227.1 12 - 13 3302 68
NM_001273299.1 NP_001260228.1 11 - 12 3236 68
NM_001273300.2 NP_001260229.2 11 - 12 3236 68
NM_001316385.1 NP_001303314.1 11 - 12 3236 68
NM_135344.6 NP_609188.4 12 - 13 3232 68
NM_164795.6 NP_723354.3 12 - 13 3458 68

View Full Product Details