The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042881.5 | NP_001036346.3 | 14 - 15 | 4760 | 69 |
NM_001169436.3 | NP_001162907.1 | 13 - 14 | 4538 | 69 | |
NM_001201789.2 | NP_001188718.1 | 14 - 15 | 4595 | 69 | |
NM_001201790.2 | NP_001188719.1 | 13 - 14 | 4538 | 69 | |
NM_001273297.1 | NP_001260226.1 | 15 - 16 | 4817 | 69 | |
NM_001273298.1 | NP_001260227.1 | 14 - 15 | 4604 | 69 | |
NM_001273299.1 | NP_001260228.1 | 13 - 14 | 4538 | 69 | |
NM_001273300.2 | NP_001260229.2 | 13 - 14 | 4538 | 69 | |
NM_001316385.1 | NP_001303314.1 | 13 - 14 | 4538 | 69 | |
NM_135344.6 | NP_609188.4 | 14 - 15 | 4534 | 69 | |
NM_164795.6 | NP_723354.3 | 14 - 15 | 4760 | 69 |