Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Piezo
Entrez Gene ID
34112
Gene Name
CG44122-PE
Gene Aliases
CG18103, CG31608, CG44122, CG8486, DmPIEZO, DmPiezo, Dmel\CG44122, Dmel_CG44122, Dmpiezo, OrfKD, anon-EST:Liang-1.32, clone 1.32, fos28F, fos500, piezo
Chromosome Location
Chr.2L: 8163073 - 8190137 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264953
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042881.5 NP_001036346.3 26 - 27 6812 63
NM_001169436.3 NP_001162907.1 26 - 27 6857 63
NM_001201789.2 NP_001188718.1 25 - 26 6509 63
NM_001201790.2 NP_001188719.1 24 - 25 6452 63
NM_001273297.1 NP_001260226.1 25 - 26 6512 63
NM_001273298.1 NP_001260227.1 24 - 25 6452 63
NM_001273299.1 NP_001260228.1 23 - 24 6398 63
NM_001273300.2 NP_001260229.2 25 - 26 7313 63
NM_001316385.1 NP_001303314.1 27 - 28 7718 63
NM_135344.6 NP_609188.4 26 - 27 6601 63
NM_164795.6 NP_723354.3 26 - 27 6926 63

View Full Product Details