Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
alien
Entrez Gene ID
34225
Gene Name
CG9556-PA
Gene Aliases
Alien, BcDNA:LD10463, CG9556, CH2, CSN2, Csn2, DCH2, Dch2, Dmel\CG9556, Dmel_CG9556
Chromosome Location
Chr.2L: 8993818 - 8997379 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013746
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273347.1 NP_001260276.1 2 - 3 369 69
NM_001298842.1 NP_001285771.1 2 - 3 369 69
NM_078793.6 NP_523517.1 2 - 3 369 69
NM_164843.3 NP_723438.1 2 - 3 369 69
GenBank mRNA AF129079.1 - 2 - 3 329 69

View Full Product Details