Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nckx30C
Entrez Gene ID
45248
Gene Name
CG18660-PA
Gene Aliases
CG18660, CG4106, Dmel\CG18660, Dmel_CG18660, NcKX30C, NckX30C, Nckx30c, potassium-dependent sodium/calcium
Chromosome Location
Chr.2L: 9711512 - 9746495 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028704
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273367.1 NP_001260296.1 5 - 6 2943 69
NM_164869.3 NP_723482.1 5 - 6 2970 69
NM_164870.3 NP_723483.1 5 - 6 2943 69

View Full Product Details