Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RluA-1
Entrez Gene ID
34438
Gene Name
RluA pseudouridine synthase 1
Gene Aliases
CG13143, CG31719, CG7475, Dmel\CG31719, Dmel_CG31719, RluA1
Chromosome Location
Chr.2L: 10463340 - 10474349 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051719
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273422.1 NP_001260351.1 3 - 4 904 81
NM_164930.2 NP_723592.1 3 - 4 1174 81
NM_164931.2 NP_723593.1 3 - 4 1308 81
NM_164932.2 NP_723594.1 3 - 4 1015 81
GenBank mRNA BT044578.1 - 3 - 4 1173 81

View Full Product Details