Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JhI-21
Entrez Gene ID
34624
Gene Name
Juvenile hormone Inducible-21
Gene Aliases
CG12317, Dmel\CG12317, Dmel_CG12317, JHI-21, Jhl-21
Chromosome Location
Chr.2L: 12051755 - 12056042 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028425
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259067.1 NP_001245996.1 6 - 7 1272 74
NM_001298934.1 NP_001285863.1 6 - 7 1242 74
NM_001298935.1 NP_001285864.1 6 - 7 1305 74
NM_135698.4 NP_609542.1 6 - 7 1248 74
NM_164988.4 NP_723717.2 6 - 7 1212 74
GenBank mRNA AY051907.1 - 6 - 7 1197 74

View Full Product Details