Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jon25Bii
Entrez Gene ID
33707
Gene Name
Jonah 25Bii
Gene Aliases
25B, CG8869, Dmel\CG8869, Dmel_CG8869, Jon25, Jon25B, SP112
Chromosome Location
Chr.2L: 4952243 - 4953136 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031654
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298679.1 NP_001285608.1 1 - 1 129 77
NM_135038.2 NP_608882.1 1 - 1 129 77

View Full Product Details