Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elgi
Entrez Gene ID
39735
Gene Name
early girl
Gene Aliases
CG17033, Dmel\CG17033, Dmel_CG17033, Nrdp1, dNrdp1
Chromosome Location
Chr.3L: 15956550 - 15958433 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283649
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274975.1 NP_001261904.1 1 - 2 214 64
NM_140559.1 NP_648816.1 1 - 2 291 64
GenBank mRNA AY051931.1 - 1 - 2 215 64

View Full Product Details