Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDCD-5
Entrez Gene ID
39776
Gene Name
Programmed Cell Death 5
Gene Aliases
BcDNA:RE17056, CG13072, DMPDCD-5, Dmel\CG13072, Dmel_CG13072, PDCD5
Chromosome Location
Chr.3L: 16231081 - 16232038 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036580
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_140591.4 NP_648848.1 1 - 2 205 62
GenBank mRNA AF320758.1 - 1 - 2 69 62
AY071119.1 - 1 - 2 199 62
KX531626.1 - 1 - 2 69 62

View Full Product Details