Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Lasp
- Entrez Gene ID
-
39864
- Gene Name
- CG3849-PA
- Gene Aliases
-
CG3849, Dmel\CG3849, Dmel_CG3849, anon-73Bd, cDNA B, lasp
- Chromosome Location
-
Chr.3L: 16676406 - 16711677 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0063485
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140655.3 |
NP_648912.2 |
4 - 5 |
1100 |
73 |
GenBank mRNA |
BT003801.1 |
- |
4 - 5 |
655 |
73 |
View Full Product Details