Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Edc3
- Entrez Gene ID
-
39957
- Gene Name
- Enhancer of decapping 3
- Gene Aliases
-
CG6311, DmEDC3, Dmel\CG6311, Dmel_CG6311, EDC3, LSM16, LSm16
- Chromosome Location
-
Chr.3L: 17525333 - 17529861 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036735
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140735.3 |
NP_648992.1 |
3 - 4 |
2031 |
92 |
GenBank mRNA |
BT099563.1 |
- |
3 - 4 |
2009 |
92 |
View Full Product Details