Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nufip
Entrez Gene ID
40043
Gene Name
CG4076-PA
Gene Aliases
CG4076, Dmel\CG4076, Dmel_CG4076, NUFIP
Chromosome Location
Chr.3L: 18746611 - 18748367 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036812
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_140801.4 NP_649058.1 1 - 2 254 82
GenBank mRNA AF429303.1 - 1 - 2 212 82

View Full Product Details