Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rad9
Entrez Gene ID
40054
Gene Name
CG3945-PA
Gene Aliases
CG3945, DmRad9, Dmel\CG3945, Dmel_CG3945, rad9
Chromosome Location
Chr.3L: 18870045 - 18872553 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025807
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104169.3 NP_001097639.2 2 - 3 326 63
NM_140809.3 NP_649066.1 2 - 3 326 63
GenBank mRNA AF076846.1 - 2 - 3 293 63
AF124502.1 - 2 - 3 294 63
AY061116.1 - 2 - 3 293 63

View Full Product Details