Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir75d
- Entrez Gene ID
-
40065
- Gene Name
- Ionotropic receptor 75d
- Gene Aliases
-
CG14076, CT33663, DmelIR75d, Dmel\CG14076, Dmel_CG14076, IR75d, lr75d
- Chromosome Location
-
Chr.3L: 18908260 - 18911182 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036829
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140817.3 |
NP_649074.2 |
3 - 4 |
1780 |
97 |
- Gene Symbol
- CG14077
- Entrez Gene ID
-
40066
- Gene Name
- CG14077-PB
- Gene Aliases
-
Dmel\CG14077, Dmel_CG14077, VIa
- Chromosome Location
-
Chr.3L: 18908260 - 18914150 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036830
View Full Product Details