Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Grasp65
- Entrez Gene ID
-
40177
- Gene Name
- CG7809-PA
- Gene Aliases
-
CG7809, Dmel\CG7809, Dmel_CG7809, GRASP, GRASP65, dGRASP, dGrasp65, dgrasp
- Chromosome Location
-
Chr.3L: 19924943 - 19927175 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036919
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140903.2 |
NP_649160.1 |
1 - 2 |
168 |
72 |
View Full Product Details