Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wb
Entrez Gene ID
43946
Gene Name
wing blister
Gene Aliases
&agr1, 2, 2Lan, 2]], A1, BEST:CK02229, BG:DS03792.1, BG:DS03792.2, CG15288, CG32970, CG42677, CK02229, CT35236, DLAM, Dmel\CG42677, Dmel_CG42677, LM-A/alpha2, LanA1, Wb, alpha[[1, fus5, l(2)09437, l(2)34Fb, l(2)br1, l34Fb
Chromosome Location
Chr.2L: 14263027 - 14328262 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261563
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057442.5 NP_476790.3 2 - 3 1436 86
NM_165086.3 NP_723870.1 2 - 3 1436 86

View Full Product Details