Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or35a
- Entrez Gene ID
-
34918
- Gene Name
- Odorant receptor 35a
- Gene Aliases
-
35a, 36E.1, BACR44L22.5, BG:BACR44L22.5, CG17868, DOR91, Dmel\CG17868, Dmel_CG17868, OR35a
- Chromosome Location
-
Chr.2L: 15622068 - 15623558 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028946
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165117.2 |
NP_723916.1 |
2 - 3 |
982 |
77 |
View Full Product Details