Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- VhaSFD
- Entrez Gene ID
-
34997
- Gene Name
- Vacuolar H[+]-ATPase SFD subunit
- Gene Aliases
-
144191_at, ATP6V1H, CG17332, Dmel\CG17332, Dmel_CG17332, V-ATPase, V-type H+-ATPase, V1H, VA, vhaSFD
- Chromosome Location
-
Chr.2L: 16722883 - 16727826 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027779
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165177.2 |
NP_723992.1 |
2 - 3 |
673 |
88 |
View Full Product Details