Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ucp4C
- Entrez Gene ID
-
33832
- Gene Name
- CG9064-PA
- Gene Aliases
-
CG9064, DmUCP4C, Dmel\CG9064, Dmel_CG9064, UCP4C, UCP4c, Ucp4c, anon-WO02079478.1, anon-WO02079478.2, anon-WO0242455.3, anon-WO0242455.4, anon-WO03037362.7, anon-WO03061681.1, anon-WO03061681.2, dmUCP4c, ucp4C
- Chromosome Location
-
Chr.2L: 5968566 - 5969761 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031757
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135132.3 |
NP_608976.1 |
1 - 1 |
1084 |
62 |
GenBank mRNA |
AY089345.1 |
- |
1 - 1 |
1053 |
62 |
View Full Product Details