Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ppk14
- Entrez Gene ID
-
33887
- Gene Name
- pickpocket 14
- Gene Aliases
-
CG9501, Dmel\CG9501, Dmel_CG9501, PPK14
- Chromosome Location
-
Chr.2L: 6354956 - 6356672 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031803
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135173.2 |
NP_609017.2 |
1 - 2 |
202 |
95 |
View Full Product Details