Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Piezo
Entrez Gene ID
34112
Gene Name
CG44122-PE
Gene Aliases
CG18103, CG31608, CG44122, CG8486, DmPIEZO, DmPiezo, Dmel\CG44122, Dmel_CG44122, Dmpiezo, OrfKD, anon-EST:Liang-1.32, clone 1.32, fos28F, fos500, piezo
Chromosome Location
Chr.2L: 8163073 - 8190137 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264953
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169436.3 NP_001162907.1 21 - 22 5911 83
NM_001316385.1 NP_001303314.1 22 - 23 6772 83
NM_135344.6 NP_609188.4 21 - 22 5655 83

View Full Product Details