Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Grip75
Entrez Gene ID
34441
Gene Name
CG6176-PA
Gene Aliases
75p, CG6176, DGrip75, Dgrip75, Dgrip75/d75p, Dgrip75s, Dmel\CG6176, Dmel_CG6176, d75p, dGrip75, grip75
Chromosome Location
Chr.2L: 10483295 - 10485857 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026431
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135568.4 NP_609412.1 1 - 2 130 67
GenBank mRNA AJ249678.1 - 1 - 2 107 67
BT150219.1 - 1 - 2 126 67

View Full Product Details