Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCAR
Entrez Gene ID
34519
Gene Name
CG4636-PA
Gene Aliases
BEST:SD02991, CG4636, D-SCAR, DWave, DmSCAR, Dmel\CG4636, Dmel_CG4636, Dscar, SCAR/WAVE, Scar, WAVE, WAVE/SCAR, Wave, dWAVE, l(2)k03107, l(2)k13811, scar, wave
Chromosome Location
Chr.2L: 10976772 - 10982395 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041781
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259037.3 NP_001245966.1 5 - 6 2003 83
NM_001259038.3 NP_001245967.1 5 - 6 2000 83
NM_001273439.2 NP_001260368.1 6 - 7 2204 83
NM_001273440.2 NP_001260369.1 4 - 5 1919 83
NM_001298895.1 NP_001285824.1 5 - 6 2123 83
NM_135633.4 NP_609477.1 4 - 5 1919 83
GenBank mRNA BT126168.1 - 5 - 6 1910 83
BT133231.1 - 4 - 5 1811 83

View Full Product Details