Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nrv3
Entrez Gene ID
35408
Gene Name
nervana 3
Gene Aliases
CG8663, Dmel\CG8663, Dmel_CG8663, JYbeta1, Nrv3, nervana3
Chromosome Location
Chr.2L: 21380000 - 21393319 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032946
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136264.6 NP_610108.1 4 - 5 780 82
NM_165378.1 NP_724339.1 4 - 5 936 82
NM_165379.3 NP_724340.1 4 - 5 803 82
GenBank mRNA AF202633.1 - 4 - 5 700 82
AY314744.1 - 4 - 5 707 82
BT044501.1 - 4 - 5 936 82

View Full Product Details