Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tdc2
- Entrez Gene ID
-
246620
- Gene Name
- Tyrosine decarboxylase 2
- Gene Aliases
-
CG30446, CG3686, Dmel\CG30446, Dmel_CG30446, TDC, TDC2, TdC2, Tdc, dTdc2, tdc, tdc2
- Chromosome Location
-
Chr.2R: 6684745 - 6689974 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0050446
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165483.3 |
NP_724489.1 |
4 - 5 |
1035 |
70 |
GenBank mRNA |
BT083414.1 |
- |
4 - 5 |
1019 |
70 |
View Full Product Details