Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scra
Entrez Gene ID
35696
Gene Name
scraps
Gene Aliases
ABP8, Abp8, Ani, Anil, Anillin, CG2092, Dmel\CG2092, Dmel_CG2092, Scra, ani, anil, anillin, l(2)03427, l(2)43Ec, l(2)k08255
Chromosome Location
Chr.2R: 7590119 - 7595242 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261385
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_165543.2 NP_724582.1 5 - 6 2125 82
NM_165544.2 NP_724583.1 6 - 7 1961 82
GenBank mRNA AY069520.1 - 5 - 6 1981 82

View Full Product Details