Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trpl
Entrez Gene ID
36003
Gene Name
transient receptor potential-like
Gene Aliases
CG1694, CG18345, DmTRPL, Dmel\CG18345, Dmel_CG18345, TRP-Like, TRPL, TrpL, Trpl, dTRPL, dTrpL, trp-l, trpL
Chromosome Location
Chr.2R: 9753697 - 9765518 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005614
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057547.5 NP_476895.1 8 - 9 1715 70
NM_165694.3 NP_724822.1 8 - 9 1840 70
NM_165695.2 NP_724823.2 8 - 9 1702 70
GenBank mRNA BT099607.1 - 8 - 9 1686 70

View Full Product Details