Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bys
Entrez Gene ID
31701
Gene Name
by S6
Gene Aliases
CG1430, Dmel\CG1430, Dmel_CG1430
Chromosome Location
Chr.X: 7901880 - 7903643 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010292
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078519.3 NP_511074.1 4 - 5 1272 68
GenBank mRNA AY061176.1 - 4 - 5 1250 68

View Full Product Details