Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- sws
- Entrez Gene ID
-
31716
- Gene Name
- swiss cheese
- Gene Aliases
-
CG2212, Dmel\CG2212, Dmel_CG2212, PNPLA6, SWS, Sws, olfE
- Chromosome Location
-
Chr.X: 7956820 - 7968236 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003656
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078520.3 |
NP_511075.3 |
7 - 8 |
3919 |
59 |
View Full Product Details