Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fw
Entrez Gene ID
32162
Gene Name
furrowed
Gene Aliases
CG1500, CT3745, Dmel\CG1500, Dmel_CG1500, fw-like, wr
Chromosome Location
Chr.X: 11992915 - 12003914 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001083
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103491.1 NP_001096961.1 6 - 7 1509 57
NM_001298217.1 NP_001285146.1 7 - 8 1889 57
NM_078581.3 NP_511136.2 8 - 9 2269 57
GenBank mRNA BT003214.1 - 8 - 9 2266 57

View Full Product Details