Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NetA
Entrez Gene ID
32398
Gene Name
Netrin-A
Gene Aliases
CG18657, CT27014, Dmel\CG18657, Dmel_CG18657, NetrinA, net, netA, netrin, netrin A
Chromosome Location
Chr.X: 14604012 - 14653928 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015773
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258738.2 NP_001245667.1 6 - 7 2637 84
NM_001298314.1 NP_001285243.1 6 - 7 2727 84
NM_078599.5 NP_511154.3 6 - 7 2633 84
GenBank mRNA AY089580.1 - 6 - 7 2634 84

View Full Product Details