Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Flo2
Entrez Gene ID
32425
Gene Name
Flotillin 2
Gene Aliases
CG11547, CG14409, CG32593, DmFlo2, Dmel\CG32593, Dmel_CG32593, FLO2, FLO[Dm]-2, Flo, Flo-2, flo2, mdcds_54791, reggie-1
Chromosome Location
Chr.X: 14839376 - 14933946 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264078
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169287.2 NP_001162758.1 3 - 4 1037 107
NM_001169288.1 NP_001162759.1 2 - 3 704 107
NM_001272623.2 NP_001259552.1 5 - 6 1085 107
NM_001272624.2 NP_001259553.1 6 - 7 1124 107
NM_001272625.2 NP_001259554.1 5 - 6 1055 107
NM_001272626.2 NP_001259555.1 6 - 7 1435 107
NM_078602.4 NP_511157.2 5 - 6 1055 107
NM_167415.3 NP_727797.1 6 - 7 1094 107
NM_167416.3 NP_727798.1 6 - 7 1206 107
NM_167417.3 NP_727799.1 5 - 6 1167 107
NM_167419.2 NP_727812.2 2 - 3 1447 107
NM_167421.2 NP_727814.1 2 - 3 1017 107
GenBank mRNA BT009962.1 - 5 - 6 1053 107
BT099913.1 - 2 - 3 1017 107
BT126133.1 - 6 - 7 1073 107

View Full Product Details