Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eag
Entrez Gene ID
32428
Gene Name
ether a go-go
Gene Aliases
CG10952, CIKE, Dmel\CG10952, Dmel_CG10952, EAG, Eag
Chromosome Location
Chr.X: 14955477 - 15009989 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000535
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042810.3 NP_001036275.1 7 - 8 1458 83
NM_001201662.2 NP_001188591.1 6 - 7 1407 83
NM_001201663.2 NP_001188592.1 7 - 8 1452 83
NM_001272629.1 NP_001259558.1 6 - 7 1407 83
NM_078603.4 NP_511158.2 7 - 8 1458 83
GenBank mRNA BT150281.1 - 4 - 5 857 83

View Full Product Details