Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eag
Entrez Gene ID
32428
Gene Name
ether a go-go
Gene Aliases
CG10952, CIKE, Dmel\CG10952, Dmel_CG10952, EAG, Eag
Chromosome Location
Chr.X: 14955477 - 15009989 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000535
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042810.3 NP_001036275.1 11 - 12 2165 81
NM_001201662.2 NP_001188591.1 10 - 11 2114 81
NM_001201663.2 NP_001188592.1 11 - 12 2159 81
NM_001272627.1 NP_001259556.1 10 - 11 2108 81
NM_001272628.1 NP_001259557.1 10 - 11 2135 81
NM_001272629.1 NP_001259558.1 10 - 11 2114 81
NM_078603.4 NP_511158.2 11 - 12 2165 81
GenBank mRNA BT150281.1 - 8 - 9 1564 81

View Full Product Details