Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mei-41
- Entrez Gene ID
-
32608
- Gene Name
- meiotic 41
- Gene Aliases
-
4252, ATR, CG4252, DmATR, Dmel\CG4252, Dmel_CG4252, Dmmei41, MEI41, MEI41/FRP1, Mei-41, Mei-41/DATR, Mei41, atr, dATR, fs(1)M37, mei-195, mei41, meiP41, mus(1)103, mus(1)104, mus-103, mus-104, mus103, mus104
- Chromosome Location
-
Chr.X: 16391698 - 16399850 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004367
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078645.3 |
NP_523369.2 |
4 - 5 |
7485 |
92 |
GenBank mRNA |
AY282465.1 |
- |
4 - 5 |
7485 |
92 |
View Full Product Details