Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mst
Entrez Gene ID
33119
Gene Name
misato
Gene Aliases
CG1424, Dmel\CG1424, Dmel_CG1424, LB-20, LB20, l(1)19Fg, l(1)LB14, l(1)LB20, lLB14
Chromosome Location
Chr.X: 21388977 - 21391533 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020272
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298589.1 NP_001285518.1 1 - 2 114 60
NM_078711.5 NP_523435.1 1 - 2 114 60

View Full Product Details