Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or22a
- Entrez Gene ID
-
33335
- Gene Name
- Odorant receptor 22a
- Gene Aliases
-
22A.1, 22a, AN11, CG12193, DOR22A.1, DOR22a, DOR53, DmOr22a, Dmel Or22a, Dmel22a, Dmel\CG12193, Dmel_CG12193, Dor53, OR22a, Or22, Or22A.1, Or22a/b, Or53, dOr22a, dor53
- Chromosome Location
-
Chr.2L: 1520491 - 1522156 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026398
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078729.3 |
NP_523453.1 |
1 - 2 |
253 |
82 |
View Full Product Details