Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daw
Entrez Gene ID
33474
Gene Name
dawdle
Gene Aliases
ALP23B, Alp, Alp23B, Alp23b, CG16987, DAW, Daw, Dmel\CG16987, Dmel_CG16987, cg16987
Chromosome Location
Chr.2L: 2805261 - 2812376 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031461
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273038.1 NP_001259967.1 2 - 3 1614 66
NM_078737.3 NP_523461.1 2 - 3 1488 66
NM_164503.2 NP_722840.1 2 - 3 1614 66
GenBank mRNA AY051485.1 - 2 - 3 1487 66

View Full Product Details