Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- gkt
- Entrez Gene ID
-
33530
- Gene Name
- glaikit
- Gene Aliases
-
CG8825, CG8826, Dmel\CG8825, Dmel_CG8825, Gkt, Glaikit, TDP1, Tdp1, anon-23Db, c22
- Chromosome Location
-
Chr.2L: 3159950 - 3162399 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0260817
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078741.4 |
NP_523465.2 |
3 - 4 |
1133 |
69 |
GenBank mRNA |
AY051884.1 |
- |
3 - 4 |
1100 |
69 |
View Full Product Details