Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ed
Entrez Gene ID
33619
Gene Name
echinoid
Gene Aliases
1X5, CG12676, CG15424, CG16842, CT13476, CT35484, Dmel\CG12676, Dmel_CG12676, E(Elp)24D, E(Elp)[24D], Ed, l(2)k01102
Chromosome Location
Chr.2L: 4031377 - 4115749 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000547
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273084.1 NP_001260013.1 8 - 9 4243 68
NM_078745.3 NP_523469.2 8 - 9 4243 68

View Full Product Details